missing translation for 'onlineSavingsMsg'
Learn More

Losartan Carboxaldehyde-d3, TRC

Product Code. 30478208
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478208 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478208 Supplier Toronto Research Chemicals Supplier No. TRCL4705071MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Losartan Carboxaldehyde-d3
CAS 1184999-26-5
Molecular Formula C22 D3 H18 Cl N6 O
Quantity 1 mg
Synonym 2-(Butyl-d3)-4-chloro-1-[[2’-(1H-tetrazol-5-yl)[1,1’-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxaldehyde, EXP 3179, DUP 167
InChI Formula InChI=1S/C22H21ClN6O/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22/h4-7,9-12,14H,2-3,8,13H2,1H3,(H,25,26,27,28)/i1D3
SMILES [2H]C([2H])([2H])CCCc1nc(Cl)c(C=O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
IUPAC Name 5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]-2-(4,4,4-trideuteriobutyl)imidazole-4-carbaldehyde
Molecular Weight (g/mol) 423.91
Formula Weight 423.1654
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.