missing translation for 'onlineSavingsMsg'
Learn More

Losartan, MedChemExpress

Product Code. 16489847
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489847 5 g 5g
16461689 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489847 Supplier Medchem Express Supplier No. HY17512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Losartan is an angiotensin II receptor antagonist, competing with the binding of angiotensin II to AT1 receptors with IC50 of 20 nM.

Losartan is an angiotensin II receptor antagonist, competing with the binding of angiotensin II to AT1 receptors with IC50 of 20 nM.

Specifications

Chemical Name or Material Losartan
CAS 114798-26-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H23ClN6O
Quantity 5 g
Synonym DuP-753
Solubility Information DMSO : ≥ 100 mg/mL (236.46 mM)
SMILES OCC1=C(Cl)N=C(CCCC)N1CC2=CC=C(C3=CC=CC=C3C4=NNN=N4)C=C2
Molecular Weight (g/mol) 422.91
Formula Weight 422.91
Percent Purity 99.55%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.