missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Lovastatin

Product Code. 18064802
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064802 10 mg 10mg
18014072 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064802 Supplier R&D Systems™ Supplier No. 1530/10
  • ONLINE EXCLUSIVE PRICE
    161.65 €
    145.95 € / 10mg
    Save 15.70 €
    9% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent HMG-CoA reductase inhibitor

Potent, competitive inhibitor of HMG-CoA reductase (Ki = 0.6 nM) therefore decreases cholesterol biosynthesis, in vitro and in vivo. Indirectly inhibits Rho-GTPase activity by depleting intracellular pool of isoprene residues. Decreases CDK2, 4, 6 and cyclin E levels and induces G1 arrest and apoptosis in tumor cell lines in vitro. Inactive lactam prodrug of lovastatin hydroxy acid, naturally bioactivated in vivo following oral administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75330-75-5
Molecular Formula C24H36O5

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 404.54
Purity >98%
Molecular Weight (g/mol) 404.55
Chemical Name or Material Lovastatin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.