missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin, 97%

Product Code. 15499537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499537 25 g 25g
15489537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499537 Supplier Thermo Scientific Alfa Aesar Supplier No. H52792.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75330-75-5 | C24H36O5 | 404.55 g/mol

Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.

Solubility
Soluble in DMSO (10 mg/ml), ethanol (10 mg/ml), methanol, chloroform, acetone, acetonitrile, DMF, and water (0.4 µg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75330-75-5
Molecular Formula C24H36O5
Molecular Weight (g/mol) 404.55
MDL Number MFCD00072164
InChI Key PCZOHLXUXFIOCF-BXMDZJJMSA-N
Synonym lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin
PubChem CID 53232
ChEBI CHEBI:40303
IUPAC Name (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Melting Point 175°C
Quantity 25 g
Solubility Information Soluble in DMSO (10mg/ml),ethanol (10mg/ml),methanol,chloroform,acetone,acetonitrile,DMF,and water (0.4 μg/ml).
Formula Weight 404.56
Percent Purity 97%
Chemical Name or Material Lovastatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.