Learn More
Lovastatin, 97%, Thermo Scientific™
96.10 € - 354.00 €
Chemical Identifiers
| CAS | 75330-75-5 |
|---|---|
| Molecular Formula | C24H36O5 |
| Molecular Weight (g/mol) | 404.55 |
| MDL Number | MFCD00072164 |
| InChI Key | PCZOHLXUXFIOCF-BXMDZJJMSA-N |
| Synonym | lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin |
| PubChem CID | 53232 |
| ChEBI | CHEBI:40303 |
| IUPAC Name | (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate |
| SMILES | CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15489537
|
Thermo Scientific Alfa Aesar
H52792.06 |
5g |
96.10 €
5g |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15499537
|
Thermo Scientific Alfa Aesar
H52792.14 |
25g |
354.00 €
25g |
Estimated Shipment: 02-10-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsLovastatin is a cholesterol lowering drug and competitive inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. It increases cellular resistance to anticancer agents such as doxorubicin and induces apoptosis in myeloma cells. It causes cell cycle arrest in G1 and G2/M phases.
Solubility
Soluble in DMSO (10 mg/ml), ethanol (10 mg/ml), methanol, chloroform, acetone, acetonitrile, DMF, and water (0.4 µg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
| 75330-75-5 | |
| 404.55 | |
| PCZOHLXUXFIOCF-BXMDZJJMSA-N | |
| 53232 | |
| (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate |
| C24H36O5 | |
| MFCD00072164 | |
| lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin | |
| CHEBI:40303 | |
| CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12 |
Specifications
| 75330-75-5 | |
| MFCD00072164 | |
| Soluble in DMSO (10mg/ml),ethanol (10mg/ml),methanol,chloroform,acetone,acetonitrile,DMF,and water (0.4 μg/ml). | |
| CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12 | |
| 404.55 | |
| CHEBI:40303 | |
| 97% |
| C24H36O5 | |
| lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin | |
| PCZOHLXUXFIOCF-BXMDZJJMSA-N | |
| (1S,3R,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate | |
| 53232 | |
| 404.56 | |
| Lovastatin |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H361d
missing translation for 'rtecsNumber' : EK7907000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.