missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin, TRC

Product Code. 17904395
Change view
Click to view available options
Quantity:
5 mg
50 mg
1 g
Unit Size:
1g
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17904395 50 mg 50mg
17910965 5 mg 5mg
17964495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17904395 Supplier Toronto Research Chemicals Supplier No. TRCL47222550MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75330-75-5
Molecular Formula C24 H36 O5
Molecular Weight (g/mol) 404.54
Synonym Lovastatin, Simvastatin Imp. E (EP), Butanoic acid, 2-methyl-, 1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-(tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl)ethyl]-1-naphthalenyl ester, [1S-[1α(R*),3α,7β,8β(2S*,4S*),8aβ]]-, (+)-Mevinolin, 6α-Methylcompactin, Altocor, Altoprev, Antibiotic MB 530B, L 154803, Lestric, Lostatin, Lovacard, Lovalip, Lovastatin, Lovastatin lactone, Lovex, MK 803, MSD 803, Mevacor, Mevinacor, Mevinolin, Mevlor, Monacolin K, Monacolin K lactone, NSC 633781, Sivlor
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15-,16-,18+,19+,20-,21-,23-/m0/s1
Formula Weight 404.2563
Chemical Name or Material Mevinolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.