missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Alkalinity

Product Code. 10779535
Click to view available options
Range:
20 to 240mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10779535

Brand: Lovibond™ 147450

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Alkacheck Tablets (100 pack)
  • Sample Cells (2)
  • Tablet Crusher
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, alkacheck tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 20 to 240mg/L
Type Alkalinity Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.