missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine: Ultra HR: 2-18% NaOCl

Product Code. 10329414
Change view
Click to view available options
Range:
2 to 18% NaOCl
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10329414 2 to 18% NaOCl Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10329414 Supplier Lovibond™ Supplier No. 147490
  • ONLINE EXCLUSIVE PRICE
    90.85 €
    80.76 € / Pack of 1
    Save 10.09 €
    11% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Acidifying GP Tablets (100 pack)
  • Chlorine HR (KI) Tablets (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Dilution Set for sample preparation
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, acidifying GP tablets, chlorine HR (Kl) tablets, 2 x 13.5mm sample cells, tablet crusher, dilution set for sample preparation, instructions, carrying case
No. of Tests 100
Range 2 to 18% NaOCl
Type Chlorine Test Kit
compliance-icons compliance-icons
Product Identifier
  • Comparator Checkit, Natriumhypochlorit, 2- 18 % .
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium iodide, citric acid, adipic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.