missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Iron
Accurate test kits for routine water analysis
Brand: Lovibond™ 147220
Code : 04
Additional Details : Weight : 0.43300kg
Includes:
- Aqua Comparator Housing
- Aqua Comparator Disc
- Iron LR Tablets (100 pack)
- Iron (II) LR Tablets (100 pack)
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
Description
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Aqua comparator™ housing, aqua comparator™ disc, iron LR tablets, iron (II) LR tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
0 to 1mg/L |
Routine water analysis | |
100 | |
LR Iron Test Kit |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.