missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Nitrite LR

Product Code. 10484292
Change view
Click to view available options
Range:
0 to 0.5mg/L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10484292 0 to 0.5mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10484292 Supplier Lovibond™ Supplier No. 147300
  • ONLINE EXCLUSIVE PRICE
    73.40 €
    65.22 € / Pack of 1
    Save 8.18 €
    11% Off
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Comparator, color disc, sample cells, reagents, instructions and carrying case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, reagents, sample cells, instructions, carrying case
No. of Tests 100
Range 0 to 0.5mg/L
Type LR Nitrite Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.