missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Phosphate LR

Product Code. 10671603
Change view
Click to view available options
Range:
0 to 4mg/L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10671603 0 to 4mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10671603 Supplier Lovibond™ Supplier No. 147240
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Phosphate No. 1 LR Tablets, (100 pack)
  • Phosphate No. 2 LR Tablets, (100 pack)
  • 2 x 13.5 mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, phosphate No. 1 LR tablets, phosphate No. 2 LR tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 30
Range 0 to 4mg/L
Type LR Phosphate AC Test Kit
compliance-icons
Product Identifier
  • comparitor phosphate LR
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: disodium disulphite,sodium hydrogensulphate, adipic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.