missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Silica VLR
Click to view available options
Range:
0 to 1mg/L
Unit Size:
Each
Description
Includes
- Aqua Comparator Housing
- Aqua Comparator Disc
- Silica No. 1 Tablets (100 pack)
- Silica No. 2 Tablets (100 pack)
- Silica PR Tablets (100 pack)
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Specifications
| Accuracy | ±5% |
| For Use With (Application) | Routine water analysis |
| Includes | Aqua comparator™ housing, aqua comparator™ disc, silica No. 1 tablets, silica No. 2 tablets, silica PR tablets, 2 x 13.5mm, sample cells, tablet crusher, instructions, carrying case |
| No. of Tests | 100 |
| Range | 0 to 1mg/L |
| Type | VLR Silica Test Kit |
Safety and Handling
Product Identifier
- Testkit Checkit Comparator D55 Siliciumdioxid VLR
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
- MIXTURE LIST-Contains : sulphamic acid, sodium molybdate, tartaric acid, boric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction