missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Powder Pack™ Reagents for Ortho (Free) Phosphate

Product Code. 12879091
Change view
Click to view available options
Type:
Powder Pack Reagents
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
12879091 Powder Pack Reagents Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12879091 Supplier Lovibond™ Supplier No. 531550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-measured, unit dose in a convenient foil pack

  • Easy to handle
  • Easy to open
  • Easy to dispense and pour
  • Specifically formulated for use in Hach™ and other manufacturer's test kits

Specifications

Type Powder Pack Reagents
For Use With (Equipment) Orbeco and Hach colorimeters and spectrophotometers
compliance-icons compliance-icons
Product Identifier
  • vario phosphate RGT F10 ml
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: dipotassium disulphate, potassium antimony(III) oxide tartrate trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.