missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Powder Packs™ for Free Chlorine
missing translation for 'orderingAttributeHoverText'
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
Förpackningsstorlek
Pack of 100
Pack of 1000
Beskrivning
Premeasured unit dose DPD reagents for water quality testing
- For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
- Also suitable for use in Hach™ instruments and test kits
- Easy-to-open, sealed foil packets eliminate risk of cross-contamination
- Available for 5, 10, and 25mL sample sizes
- Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Uppfyllande
US EPA-accepted DPD method
Specifikationer
Specifikationer
| Certifications/Compliance | US EPA |
| Volume (Metric) | 5 mL |
| Format | Powder |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer |
| Quantity | 100 Pack |
| Range | 0 to 3.5 mg/L |
| Type | DPD Free Chlorine Reagent |
| Volume (English) | 0.16 oz. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering