missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Sulfate 4 Powder Packs™
Click to view available options
Range:
0 to 70mg/L
Unit Size:
Pack of 100
Description
Premeasured unit dose reagent for water quality testing
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| Certifications/Compliance | US EPA |
| Color | White |
| Composition | Citric Acid 50 to 60%, Barium Chloride Dihydrate 40 to 50% |
| pH Range | 2.3 |
| Format | Powder |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Range | 0 to 70mg/L |
| Storage Requirements | 20°C |
| Type | Sulfate 4 Reagent |
Safety and Handling
Product Identifier
- vario sulphate 4
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- MIXTURE LIST-Contains : citric acid, bariumchloride dihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction