missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Total Chlorine Powder Packs™

Product Code. 15341655
Change view
Click to view available options
Quantity:
100 Pack
1000 Pack
Range:
0 to 2 mg/L
0 to 3.5 mg/L
0 to 5 mg/L
Volume (English):
0.16 oz.
0.33 oz.
0.84 oz.
Volume (Metric):
5 mL
10 mL
25 mL
Unit Size:
Pack of 100
Pack of 1000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Range Volume (English) Volume (Metric) unitSize
15341655 1000 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 1000
15351645 100 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL Pack of 100
15361645 1000 Pack 0 to 3.5 mg/L 0.16 oz. 5 mL Pack of 1000
10402345 100 Pack 0 to 2 mg/L 0.33 oz. 10 mL Pack of 100
15351655 100 Pack 0 to 5 mg/L 0.84 oz. 25 mL Pack of 100
15361655 1000 Pack 0 to 5 mg/L 0.84 oz. 25 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15341655 Supplier Lovibond™ Supplier No. 530123

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Premeasured unit dose DPD reagents for water quality testing

  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

US EPA-accepted DPD method

Specifications

Certifications/Compliance US EPA
Volume (Metric) 10 mL
Format Powder
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
Quantity 1000 Pack
Range 0 to 2 mg/L
Type DPD Total Chlorine Reagent
Volume (English) 0.33 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.