missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Reactive Phosphorus Orthophosphate Reagent Set
Click to view available options
No. of Tests:
50
Unit Size:
Each
Description
Includes
Phosphate 3 Reagent (10mL sample, 50 pack), Dilution Tube (50 ct.), Deionized water (100mL)
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Includes | 10mL phosphate 3 reagent (50 pack), 50 ct. dilution tube, 100mL deionized water |
| No. of Tests | 50 |
| Range | 0.06 to 5mg/L |
| Test Method | Ascorbic Acid |
| Type | Reactive Phosphorous Reagent Set |
Safety and Handling
Product Identifier
- Vario Phosphate RGT F10 mL
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- MIXTURE LIST-Contains : dipotassium disulfate, potassium antimony(III) oxide tartrate trihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction