missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, High Range

Product Code. 15208103
Click to view available options
Range:
0 to 150mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15208103

Brand: Lovibond™ 535560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide HR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL

Premeasured unit dose reagent for water quality testing

  • Offers same high quality standards as tablet reagents
  • Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open package
  • Easily create “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Specifications

For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor
Includes Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide HR tubes, 50 ct. total nitrogen Acid LR/HR tubes, 100mL deionized water
No. of Tests 25
Range 0 to 150mg/L
Test Method Persulfate Digestion
Type High Range Total Nitrogen Reagent Set
compliance-icons compliance-icons
Product Identifier
  • Vario Total Nitrogen Acid LR/HR Tube
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.