missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Phosphate No. 1 LR Tablets

Product Code. 15612514
Click to view available options
Range:
0.05 to 4mg/L
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15612514

Brand: Lovibond™ 513040BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 1 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%, Format: Tablet

Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 1 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%, Format: Tablet

Specifications

Color White
Composition Sodium Bisulfate 50 to 60%, Adipic Acid 10 to 20%, Boric Acid 25 to 35%, Citric Acid 0.1 to 1%, Potassium Antimony Oxide Tartarate Hemi hydrate <0.1%
pH Range 1.13
Format Tablet
For Use With (Application) Low range phosphate AC test kit, phosphate, ortho LR reagent set, water analysis
For Use With (Equipment) MC500 colorimeters and SP600 spectrophotometers
Range 0.05 to 4mg/L
Storage Requirements 20°C
Test Method Phosphomolybdic Acid/Ascorbic Acid
Type Phosphate No. 1 LR Tablet
compliance-icons
Product Identifier
  • phosphate No. 1 LR
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydrogen sulphate, adipic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.