missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Low-Density Polyethylene Bags

Product Code. 11878631 Shop All Cole-Parmer Products
Change view
Click to view available options
Dimensions (L x W):
127 x 76mm
178 x 127mm
229 x 152mm
254 x 203mm
305 x 254mm
457 x 152mm
457 x 305mm
Unit Size:
Each
Pack of 1000
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11878631 229 x 152mm Pack of 500
11729859 178 x 127mm Pack of 1000
11749859 305 x 254mm Pack of 500
11755648 457 x 305mm Pack of 500
11868631 127 x 76mm Pack of 1000
12303820 254 x 203mm Pack of 500
13415938 457 x 152mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 11878631 Supplier Cole-Parmer™ Supplier No. 0650510
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Store a variety of solids or powders in these LDPE bags. Low Density Polyethylene Bags stand up to your storage needs with a strong bottom seal.

  • Seal with heat sealer or bag ties
  • Good for general laboratory storage
  • Available in a variety of sizes and thicknesses

Specifications

Description Low-Density Bag
Color Clear
Material Polyethylene
Quantity 500 Pack
Thickness (English) 2.50 mil
Dimensions (L x W) 229 x 152mm
Length (Metric) 229 mm
Width (Metric) 152 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.