missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Low Profile Tubes and Flat Caps

Product Code. 10610754
Change view
Click to view available options
Product Type:
8-Strip Tube
Low Profile Tube and Flat Cap
Color:
Natural
Unit Size:
Pack of 120
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Unit Size
10610754 Low Profile Tube and Flat Cap Natural Pack of 250
10374463 8-Strip Tube Natural Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10610754 Supplier Thermo Scientific™ Supplier No. AB0776
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase the efficiency of reactions with these low-profile strip tubes for PCR and qPCR applications.

Ideal for applications requiring low reaction volumes, Thermo Scientific™ Low Profile Strip Tubes are compatible with 0.2mL cycler blocks.

The low profile design of the Thermo Scientific Low Profile Strip Tubes reduces the dead space between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Thermo Scientific low profile products are especially recommended for use with reaction volumes below 20 μL.

The low profile design of the Thermo Scientific strip tubes without caps reduces the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. Low profile products are especially recommended for use with reaction volumes below 20 μL.

Highlights

  • Suitable for 0.2 mL thermal cycler blocks
  • 150 μL maximum tube capacity when capped
  • Low profile to reduce dead space and increase PCR efficiency
  • Labelled A-H end tabs (8-strip) and 1-12 end tabs (12-strip)
Applications
  • PCR
  • qPCR/Real-Time PCR

Hazardous: No
Storage Condition: Ambient

PCR, qPCR and Real-Time PCR.

Certifications

Certified free from human DNA, RNases, and DNases.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified DNA/RNase Free
Color Natural
Volume (Metric) Working 0.1 mL
Volume (Metric) 0.15 mL
Quantity 250 strips
For Use With (Equipment) Thermal Cyclers
Material Polypropylene
Reaction Speed Low profile
Closure Type Flat Cap
Closure Included Flat Caps
Format 250 Strip Tubes and Caps
Product Type Low Profile Tube and Flat Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.