Learn More
Corning™ LSE Digital Microplate Shaker
Product Code. 11971991 Shop All Corning Products
Change view
Click to view available options
Model:
6781-4
6782-4
Unit Size:
Each
Product Code. Model unitSize
11971991 6782-4 Each
10713042 6781-4 Each
2 options
Product Code. 11971991
Brand: Corning™ 67824
Only null left
Add to basket
Add to basket
Large capacity – process up to 4 microplates
The Corning LSE digital microplate shaker features digital control of shaking speed and run time. A single control knob is used to set parameters, and values are shown on a large 3-digit display. A sturdy base encloses the motor for quiet, vibration-free operation.
The included stainless steel platform accommodates up to 4 microplates (standard or deep well), and the shaker is safe for use in temperature controlled environments from 4°C to 65°C.
  • Large capacity – process up to 4 microplates
  • 3 mm orbit for resuspending, homogenizing, or vortexing
  • Large digital display with single-knob control of speed time
  • Suitable for cold room or incubator use
TRUSTED_SUSTAINABILITY
Specifications
Model 6782-4