Learn More
Corning™ LSE Digital Microplate Shaker
Large capacity – process up to 4 microplates
1084.25 €
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
10713042
|
Corning™
6781-4 |
6781-4 |
1084.25 €
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
11971991
|
Corning™
6782-4 |
6782-4 |
1084.25 €
Each |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
Description
The Corning LSE digital microplate shaker features digital control of shaking speed and run time. A single control knob is used to set parameters, and values are shown on a large 3-digit display. A sturdy base encloses the motor for quiet, vibration-free operation.
The included stainless steel platform accommodates up to 4 microplates (standard or deep well), and the shaker is safe for use in temperature controlled environments from 4°C to 65°C.
- Large capacity – process up to 4 microplates
- 3 mm orbit for resuspending, homogenizing, or vortexing
- Large digital display with single-knob control of speed time
- Suitable for cold room or incubator use