Learn More
Tocris Bioscience™ LSN 2463359
GREENER_CHOICE
Potent and selective positive allosteric modulator at mGlu5
Brand:  Tocris Bioscience™ 5275/50
Encompass
Product Code. 15756646
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
LSN 2463359 is a potent and selective positive allosteric modulator at the mGlu5 receptor (EC50 = 24 nM). Has no activity at mGlu1-4, mGlu8 and GABAB receptors. Reverses methylazoxymethanol acetate (MAM)-induced deficits in reversal learning memory and normalizes SDZ 220-581 and RO 63-1908 -induced alterations in instrumental responses in rat models. Promotes wakefulness in vivo. Brain penetrant and orally active.
Specifications
Specifications
>99%