Learn More
IKA™ LT 5.28 Pressure Control Valve
Product Code. 17503626 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
Temperature Control Jacket of The Laboratory Reactor Vessel
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17503626 Temperature Control Jacket of The Laboratory Reactor Vessel Each
1 options
Product Code. 17503626 Supplier IKA™ Supplier No. 0020100227
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The pressure control valve LT 5.28 is used to regulate the pressure in the temperature control jacket of the laboratory reactor vessel.
  • The pressure control valve LT 5.28 is used to regulate the pressure in the temperature control jacket of the laboratory reactor vessel after the temperature control hoses have been disconnected.
  • The valve prevents the formation of an overpressure, which could result from a temperature change of the medium in the temperature control jacket.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Temperature Control Jacket of The Laboratory Reactor Vessel