missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LTURM 34

Product Code. 30306748
Click to view available options
:
10 mg
This item is not returnable. View return policy

Product Code. 30306748

Brand: Tocris Bioscience™ 5834/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent DNA-PK inhibitor; also increases CRISPR-Cas9 deletion efficiency

LTURM 34 is a potent DNA-PK inhibitor (IC50 = 34 nM). Exhibits 170-fold selectivity for DNA-PK over PI3-K. Attenuates proliferation of a wide range of tumor cell lines in vitro. LTURM 34 also increases CRISPR-Cas9 deletion efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-(4-Dibenzothienyll)-2-(4-morpholinyl)-4H-1,3-benzoxazin-4-one
CAS 1879887-96-3
Quantity 10 mg
Target DNA-dependent Protein Kinase Inhibitors
Molecular Formula C24H18N2O3S
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.