missing translation for 'onlineSavingsMsg'
Learn More

Lubiprostone, TRC

Product Code. 30137031
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137031 50 mg 50mg
30137680 1 mg 1mg
30007910 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137031 Supplier Toronto Research Chemicals Supplier No. TRCL47350050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136790-76-6
Molecular Formula C20 H32 F2 O5
Molecular Weight (g/mol) 390.46
Synonym (11α)-16,16-difluoro-11-hydroxy-9,15-dioxo-Prostan-1-oic acid, Lubiprostone, Ru 0211
IUPAC Name 7-[(1R,2R,3R)-2-(4,4-difluoro-3-oxooctyl)-3-hydroxy-5-oxocyclopentyl]heptanoic acid
SMILES CCCCC(F)(F)C(=O)CC[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H32F2O5/c1-2-3-12-20(21,22)18(25)11-10-15-14(16(23)13-17(15)24)8-6-4-5-7-9-19(26)27/h14-15,17,24H,2-13H2,1H3,(H,26,27)/t14-,15-,17-/m1/s1
Formula Weight 390.2218
Chemical Name or Material Lubiprostone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.