missing translation for 'onlineSavingsMsg'
Learn More

Lubrol, MP Biomedicals

Product Code. 11455252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455252 100 g 100g
11465252 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455252 Supplier MP Biomedicals Supplier No. 0219529980
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula HO(CH2CH2O)n C16H33 n = ∽23
Synonym Polyethylene glycolmonocetyl ether

Specifications

Color White
Physical Form Waxy Solid
Quantity 100 g
Chemical Name or Material Lubrol, Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.