Learn More
Value Plastics Luer Adapter Fittings, Tee, Polypropylene
Product Code. p-200105016 Shop All Cole-Parmer Products
Change view
Click to view available options
Width (English):
29/32 in.
37/64 in.
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) unitSize
30053986 37/64 in. Pack of 25
30052972 29/32 in. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30053986 Supplier Value Plastics Supplier No. 5012011
Add to basket
Add to basket
Easily transition to a luer fitting with directional flexibility
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards. Constructed of ADCF polypropylene
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Packaging Quantity 25/Pk.
Inlet Female Luer Thread, Female Luer Thread
Width (English) 37/64 in.