Learn More
Hamilton™ Luer to Small Hub RN Adaptor
Product Code. 12376239
Change view
Click to view available options
Product Type:
Adapter
Unit Size:
Each
Product Code. Product Type Unit Size
12376239 Adapter Each
1 options
Product Code. 12376239 Supplier Hamilton™ Supplier No. 5575301
  • ONLINE EXCLUSIVE PRICE
    67.40 €
    62.29 € / Pack of 1
    Save 5.11 €
    7% Off
View all promotions
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Small Hub Removable (RN) needles can be used with removable (RN)-style syringes that are 100 μL and smaller. Standard needle length is 2 in. / 51 mm. Removable (RN) needles are available in convenient six packs. Custom lengths and point styles are available.
Specifications
Product Type Adapter