missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Lumat LB 9510 Tube Luminometer

Product Code. 31777295
Change view
Click to view available options
Model:
High Sensitivity
Immunoassay
Reporter Gene
Standard
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
31777295 Standard Each
31777289 Reporter Gene Each
31777331 High Sensitivity Each
31777419 Immunoassay Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31777295 Supplier Berthold Technologies Supplier No. 7145010
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tube luminometer designed to support basic luminescence technologies for research applications.

  • True single photon counting combined with a low-noise photomultiplier tube for up to 6 decades of linear dynamic range
  • Excellent luminescence sensitivity: <1 zmol firefly luciferase and <1 amol ATP/tube (high sensitivity model) can be reliably detected
  • Can be equipped with up to 2 JET injectors with variable volumes and ultra-fast injection speed, eliminating delay between injection and measurement
  • JET injectors use Teflon bellows for accurate and fast injections, ensuring efficient mixing, cell-friendliness, and extreme longevity
  • LightCompass™ software for flexible creation of measurement protocols, curve fitting, qualitative analysis, and compliance with FDA 21 CFR Part 11

Specifications

Type Luminometer
Display Type No Display
Connection Type USB
Detector Low-noise Photomultiplier Tube in Single Photon Counting Mode
Detection Method Luminescence
Detection Limits <5 amol ATP
Injection Type JET Injectors
Color Light Gray
For Use With (Application) Luminescence Assays
Voltage 110/240 V
Frequency 50/60 Hz
For Use With (Equipment) Tubes
Height (Metric) 220 mm
Width (Metric) 240 mm
Depth (Metric) 280 mm
Weight (Metric) 4 kg
Includes Luminometer, Power Brick, Accessory Set, User Manual, LightCompass™ Basic Software
System Requirements PC with Windows 10 or Newer
Storage Requirements 0°C to 40°C
Certifications/Compliance CE, NRTL
Model Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.