missing translation for 'onlineSavingsMsg'
Learn More

Lumazine, TRC

Product Code. 30478207
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478207 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478207 Supplier Toronto Research Chemicals Supplier No. TRCL47380010G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lumazine
CAS 487-21-8
Molecular Formula C6 H4 N4 O2
Quantity 10 g
Synonym 2,4(1H,3H)-Pteridinedione, 2,4(3H,8H)-Pteridinedione, 2,4-Dihydroxypteridine, 2,4-Pteridinediol, Lumazin, NSC 225113, NSC 41801, Pteridine-2,4-dione
InChI Formula InChI=1S/C6H4N4O2/c11-5-3-4(8-2-1-7-3)9-6(12)10-5/h1-2H,(H2,8,9,10,11,12)
SMILES Oc1nc(O)c2nccnc2n1
IUPAC Name pteridine-2,4-diol
Molecular Weight (g/mol) 164.12
Formula Weight 164.0334
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.