missing translation for 'onlineSavingsMsg'
Learn More

Lumicitabine, MedChemExpress

Product Code. 16491637
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491637 50 mg 50mg
16491319 1 mg 1mg
16468177 5 mg 5mg
16456056 100 mg 100mg
16425468 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16491637 Supplier Medchem Express Supplier No. HY12983A
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.

Lumicitabine (ALS-008176) is an inhibitor of the respiratory syncytial virus (RSV) polymerase.

Specifications

Chemical Name or Material Lumicitabine
CAS 1445385-02-3
Color White
Purity Grade Notes Research
Molecular Formula C18H25ClFN3O6
Quantity 50 mg
Synonym ALS-008176 ALS-8176
Solubility Information DMSO : ≥ 50 mg/mL (115.24 mM)
SMILES ClC[C@@]1(COC(C(C)C)=O)[C@@H](OC(C(C)C)=O)[C@@H](F)[C@H](N2C(N=C(N)C=C2)=O)O1
Molecular Weight (g/mol) 433.86
Formula Weight 433.86
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.