missing translation for 'onlineSavingsMsg'
Learn More

Merck Immobilon Western HRP Chemiluminescence Substrates

Product Code. 12634645 Shop All Merck Products
Change view
Click to view available options
Quantity:
500 mL
Product Type:
HRP Substrate
Sensitivity:
∼1 to 3 pg
∼400 fg
∼6 pg
Unit Size:
500mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type Sensitivity unitSize
12634645 500 mL HRP Substrate ∼400 fg Each
11790644 500 mL HRP Substrate ∼1 to 3 pg Each
11770644 500 mL HRP Substrate ∼6 pg 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12634645 Supplier Merck Supplier No. WBLUF0500
  • ONLINE EXCLUSIVE PRICE
    645.00 €
    592.68 € / Pack of 1
    Save 52.32 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Premixed, ready-to-use chemiluminescent reagents for detection of HRP-based Westerns

Horseradish peroxidase (HRP) catalyzes the oxidation of luminol in the presence of peroxide, releasing photons (chemiluminescence) that are captured on x-ray films or by CCD cameras. Most HRP substrates are plagued by quickly decaying signal and short shelf life, but Luminata substrates are formulated differently to include stabilizing reagents.

  • Consistent results with less pipetting error

The stabilizers added to Luminata substrates:

  • Prevent spontaneous oxidation of luminol by peroxide in the absence of HRP
  • Enhance the stability of both the luminol and peroxide, ensuring that Luminata substrates are stable for one year in their premixed format
  • Stabilize, strengthen, and prolong the chemiluminescence signal once HRP starts to catalyze oxidation

Three different sensitivities for a range of detection needs:

  • Immobilon Classico premixed reagent for everyday detection needs
  • Immobilon Crescendo premixed reagent for detection in the picogram range
  • Immobilon Forte premixed reagent for detection in the femtogram range
TRUSTED_SUSTAINABILITY

Specifications

Product Type HRP Substrate
For Use With (Application) Dot/Slot Blot
Quantity 500 mL
Sensitivity ∼400 fg
Content And Storage 18°C to 24°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.