missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Luminescence Test Tube LB 9517

Product Code. 31777294
Change view
Click to view available options
For Use With (Equipment):
Junior LB 9509 Portable Luminometer
Lumat LB 9510 Tube Luminometer
Sirius 2 LB 9526 Tube Luminometer
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
31777294 Lumat LB 9510 Tube Luminometer Each
31777291 Junior LB 9509 Portable Luminometer Each
31777333 Sirius 2 LB 9526 Tube Luminometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31777294 Supplier Berthold Technologies Supplier No. 5565801
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-precision test tubes for routine quality control and performance verification of tube luminometers.

  • Offers the easiest, quickest, and most reliable way to periodically verify tube luminometer performance
  • Light emission generated by temperature-stabilized green LEDs for consistent output
  • Integrated diffusers simulate the light emission properties of real samples

Specifications

Type Test Tube
Light Source LED
Color Black
For Use With (Application) QC of Tube Luminometers
Voltage 6 V
For Use With (Equipment) Lumat LB 9510 Tube Luminometer
Includes Transport Case, Charging Stand, Charger, Test Tube
Certifications/Compliance CE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.