missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Berthold Technologies Luminescence Test Tube LB 9517
Description
- Offers the easiest, quickest, and most reliable way to periodically verify tube luminometer performance
- Light emission generated by temperature-stabilized green LEDs for consistent output
- Integrated diffusers simulate the light emission properties of real samples
Specifications
Specifications
| Type | Test Tube |
| Light Source | LED |
| Color | Black |
| For Use With (Application) | QC of Tube Luminometers |
| Voltage | 6 V |
| For Use With (Equipment) | Sirius 2 LB 9526 Tube Luminometer |
| Includes | Transport Case, Charging Stand, Charger, Test Tube |
| Certifications/Compliance | CE |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?