Learn More
Abcam Luminescent ATP Detection Assay Kit
Product Code. 32120073 Shop All Abcam Products
Change view
Click to view available options
Quantity:
300 Tests
Unit Size:
300 tests
Product Code. Quantity Unit Size
32120073 300 Tests 300 tests
1 options
Product Code. 32120073 Supplier Abcam Supplier No. ab113849300Test
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Detect Adenosine-5'-triphosphate in 30m in Cell Samples with Luminescent ATP Detection Assay Kit AB113849. Cited in 284 publications. Use with Microplate reader.
Detect Adenosine-5'-triphosphate in 30m in Cell Samples with Luminescent ATP Detection Assay Kit AB113849. Cited in 284 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 300 Tests