missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Luminoskan™ Microplate Luminometer, two dispensers

Product Code. 15803547 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Dispensers:
Up to 3
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Dispensers unitSize
15803547 Up to 3 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15803547 Supplier Thermo Scientific™ Supplier No. 5300332

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Useful for a variety of luminometric research applications, the Thermo Scientific Luminoskan Microplate Luminometer is a compact and robust instrument offering plate format versatility, fast reading speeds, and up to three dispensers.

This instrument has passed validation at Promega Corporation to be certified as SkanIt software is quick and easy to learn. It offers effortless data analysis (with a comprehensive selection of built-in calculations, including blank subtraction, standard curve creation, classification, quality control, kinetic analysis) and exporting capabilities. SkanIt software is an open license platform that allows unlimited installations.

Includes:
SkanIt PC Software. Filters available upon request.

Applications include:
Reporter gene assays, immunoassays with luminescent substrates, cell proliferation and cytotoxicity assays, intracellular Ca2+ assays, ATP assays, phagocytosis assays, reactive oxygen assays, microbiological assays, enzyme assays, BRET assays, ADMEtox

TRUSTED_SUSTAINABILITY

Specifications

Compatibility Robot
Detector Type Photomultiplier Tube (PMT)
Dimensions (L x W x H) 420 x 420 x 340 mm (16.5 x 16.5 x 13.4 in.)
Interface PC Software (Thermo Scientific SkanIt Software)
Measurement Speed 18 s for 96-well Plate
Measurement Technology Luminescence
Plate Type 6 to 384-well Plates
Product Line Luminoskan
Reading Options Top and Bottom
Read-out Range Up to approx. 5000 Relative Light Units (RLU)
Shaking Orbital
Software Type SkanIt Software
Wavelength Selection Filters
No. of Dispensers Up to 3
Power Consumption 200 VA max.
Shipping Condition Room Temperature
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.