missing translation for 'onlineSavingsMsg'
Learn More

Lurasidone Hydrochloride, MedChemExpress

Product Code. 16498066
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16498066

Brand: Medchem Express HYB0032

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lurasidone (Hydrochloride) (SM-13496 (Hydrochloride)) is an antagonist of both dopamine D2 and 5-HT7 with IC50s of 1.68 and 0.495 nM, respectively. Lurasidone (Hydrochloride) (SM-13496 (Hydrochloride)) is also a partial agonist of 5-HT1A receptor with an IC50 of 6.75 nM.

Lurasidone (Hydrochloride) (SM-13496 (Hydrochloride)) is an antagonist of both dopamine D2 and 5-HT7 with IC50s of 1.68 and 0.495 nM, respectively. Lurasidone (Hydrochloride) (SM-13496 (Hydrochloride)) is also a partial agonist of 5-HT1A receptor with an IC50 of 6.75 nM.

Specifications

Chemical Name or Material Lurasidone Hydrochloride
CAS 367514-88-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H37ClN4O2S
Quantity 100 mg
Synonym SM-13496 Hydrochloride
Solubility Information DMSO : 6.67 mg/mL (12.61 mM; ultrasonic and warming and adjust pH to 3 with HCl and heat to 80°C) ∣Ethanol : 2 mg/mL (3.78 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@H]([C@H]1CC[C@@H]2C1)[C@H]2C3=O)N3C[C@@H]4CCCC[C@H]4CN(CC5)CCN5C6=NSC7=CC=CC=C76.Cl
Molecular Weight (g/mol) 529.14
Formula Weight 529.14
Percent Purity 97.75%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.