missing translation for 'onlineSavingsMsg'
Learn More

Melford Luria Broth Powder Miller's LB Broth

Product Code. 30018407
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
1 kg
5 kg
missing translation for 'unitSize'
100g
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 30018407

missing translation for 'mfr': Melford L240405000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For propagation and maintenance of E.

For propagation and maintenance of E. Coli. High Salt Grade. Molecular Biology Grade.

Dissolve 25 grams in 1 liter distilled water

Adjust the pH to 7.2

Sterilize by autoclaving for 15 minutes

Ingredients:

Casein digest of peptone: 10 g/l

Yeast: 5 g/l

Sodium Chloride: 10 g/l

Specifications

Quantity 5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.