missing translation for 'onlineSavingsMsg'
Learn More

Lutein, 90%

Product Code. 15449182 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15449182 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15449182 Supplier Thermo Scientific Acros Supplier No. 456160010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-40-2 | C40H56O2 | 568.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127-40-2
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C40H56O2
Quantity 1 g
Synonym lutein
InChI Key KBPHJBAIARWVSC-HZNSELMYSA-N
SMILES CC1=C(C(CC(C1)O)(C)C)C=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC2C(=CC(CC2(C)C)O)C)C)C
IUPAC Name (1S)-4-[(1E,3Z,5E,7Z,9E,11Z,13E,15Z,17E)-18-[(1R)-4-hydroxy-2,6,6-trimethylcyclohex-2-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaenyl]-3,5,5-trimethylcyclohex-3-en-1-ol
Molecular Weight (g/mol) 568.87
PubChem CID 102514741
Formula Weight 568.87
Percent Purity 90%
Chemical Name or Material Lutein
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.