missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Luteolin

Product Code. 18064562
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064562 10 mg 10mg
18064062 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18064562 Supplier R&D Systems™ Supplier No. 2874/10
  • ONLINE EXCLUSIVE PRICE
    146.31 €
    131.25 € / 10mg
    Save 15.06 €
    10% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Anti-inflammatory, antioxidant and free radical scavenger

Anti-inflammatory, antioxidant and free radical scavenger. Inhibits LPS-induced TNF-α, IL-6 and inducible nitric oxide production and blocks NF-κB and AP-1 activation. Also inhibits TNF-α -induced COX-2 expression. Antiproliferative and chemopreventative; inhibits proliferation of Lewis lung carcinoma cells in vivo. Inhibits influenza virus replication in vitro by preventing viral entry. Molecular docking studies also indicate binding to SARS-CoV-2 main protease (Mpro or 3CLpro).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-70-3
Molecular Formula C15H10O6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 286.24
Purity >98%
Molecular Weight (g/mol) 286.239
Chemical Name or Material Luteolin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.