missing translation for 'onlineSavingsMsg'
Learn More

Lutetium foil, 0.127mm (0.005in) thick, 99.9% (REO)

Product Code. 11365709
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365709 50 x 50 mm Each
11366587 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11365709 Supplier Thermo Scientific Alfa Aesar Supplier No. 000599.FI
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7439-94-3 | Lu | 174.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-94-3
Molecular Formula Lu
Molecular Weight (g/mol) 174.97
MDL Number MFCD00011098
InChI Key OHSVLFRHMCKCQY-UHFFFAOYSA-N
Synonym lutecium, unii-5h0doz21uj, 5h0doz21uj, foil, foil, 0.127mm 0.005in thick reo, cassiopium, lutecio, cassiopeium, atom, pieces
PubChem CID 23929
ChEBI CHEBI:33382
IUPAC Name lutetium
SMILES [Lu]

Specifications

Color Silver to Gray
Quantity 50 x 50 mm
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.78g/25x25mm
Form Foil
UN Number UN3178
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lutetium foil, 0.127mm (0.005 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.