missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Luzindole

Product Code. 18084072
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084072 50 mg 50mg
18064622 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084072 Supplier Tocris Bioscience™ Supplier No. 0877/50
  • ONLINE EXCLUSIVE PRICE
    933.00 €
    882.00 € / 50mg
    Save 51.00 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Competitive melatonin MT1/MT2 antagonist

Melatonin antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117946-91-5
Molecular Formula C19H20N2O

Specifications

Quantity 50 mg
Formula Weight 292.38
Purity >99%
Molecular Weight (g/mol) 292.38
Chemical Name or Material Luzindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.