missing translation for 'onlineSavingsMsg'
Learn More

LY 294002, TRC

Product Code. 30482794
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482794 5 mg 5mg
30480284 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482794 Supplier Toronto Research Chemicals Supplier No. TRCL4865905MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154447-36-6
Molecular Formula C19H17NO3
Molecular Weight (g/mol) 307.34
Synonym NSC 697286, SF 1101, 8-Phenyl-2-(morpholin-4-yl)-chromen-4-one, 2-(4-Morpholinyl)-8-phenyl-4H-1-benzopyran-4-one, 2-Morpholino-8-phenyl-4H-chromen-4-one
SMILES COC(=O)C[C@@H](Br)C(=O)OC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H17NO3/c21-17-13-18(20-9-11-22-12-10-20)23-19-15(7-4-8-16(17)19)14-5-2-1-3-6-14/h1-8,13H,9-12H2
Chemical Name or Material LY 294002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.