missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LY 379268 disodium salt

Description
LY 379268 disodium salt is a sodium salt of LY 379268 (Cat. No. 2453), a highly selective group II mGlu receptor agonist.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | LY 379268 disodium salt |
| Purity | >98% |
| Molecular Weight (g/mol) | 231.11 |
| Inhibitors | LY 379268 disodium salt |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?