missing translation for 'onlineSavingsMsg'
Learn More

Lycopene, >90%, MP Biomedicals™

Product Code. 11476801 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 11476801

Brand: MP Biomedicals 0219868401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 502-65-8
Molecular Formula C40H56
Molecular Weight (g/mol) 536.888
InChI Key OAIJSZIZWZSQBC-GYZMGTAESA-N
Synonym lycopene, psi,psi-carotene, all-trans-lycopene, trans-lycopene, lycopene 7, lycored, lycopene van, lycopene, all-trans, unii-sb0n2n0wv6, ccris 7925
PubChem CID 446925
ChEBI CHEBI:15948
IUPAC Name (6E,8E,10E,12E,14E,16E,18E,20E,22E,24E,26E)-2,6,10,14,19,23,27,31-octamethyldotriaconta-2,6,8,10,12,14,16,18,20,22,24,26,30-tridecaene
SMILES CC(=CCCC(=CC=CC(=CC=CC(=CC=CC=C(C)C=CC=C(C)C=CC=C(C)CCC=C(C)C)C)C)C)C

Specifications

Color Red
Quantity 1 mg
Formula Weight 536.9
Percent Purity >90%
Physical Form Crystalline Needles
Chemical Name or Material Lycopene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.