missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 488

Product Code. 16875691
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16875691 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16875691 Supplier Invitrogen™ Supplier No. L32470

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 488, is a bright-green LEL- fluorophore conjugate.

The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research. LEL-DyLight 488 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.

  • Very bright DyLight 488 conjugate detected with traditional FITC filter set, Ex/Em: 493/518 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
  • Inhibiting/eluting sugar: chitin hydrolysate
  • Blood group specificity: non-specific
  • Unconjugated lectin molecular weight: 71 KDa.
  • Immunofluorescence working concentration: 5–20 μg/mL

LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+. If a precipitate forms upon long-term storage, warm to 37°C.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Source Tomato
Conjugate DyLight 488
Content And Storage When stored at or below 2–8°C in the dark, the product is stable for 6 months after receipt.
Label Type DyLight Fluors
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins
Product Line DyLight

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.