missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Lycorine hydrochloride

Product Code. 16423856
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16423856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16423856 Supplier MedChemExpress Supplier No. HYN0289
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lycorine hydrochloride is the main active ingredient of the herbal medicine derived from Lycoris radia and is also a melanoma vasculogenic inhibitor and has anti-tumor activity. Lycorine hydrochloride effectively inhibits mitotic proliferation of Hey1B cells (IC50 of 1.2 μM).

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 323.77 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 2188-68-3
Product Type Natural Products
Purity 99.35%
Solubility DMSO : 25 mg/mL (77.22 mM∣Need ultrasonic) ∣H2O : 16.67 mg/mL (51.49 mM∣Need ultrasonic)
Target Autophagy∣Bacterial∣Virus Protease
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.