Learn More
Invitrogen™ Lyo-ready Enzymes and Reagents for Recombinase Polymerase Amplification (RPA)
Description
Invitrogen Lyo-ready recombinase polymerase amplification (RPA) reagents are designed for fast, sensitive, and specific DNA amplification through isothermal RPA reactions. Lyo-ready RPA reagents are available in stand-alone and kit formats. Reverse transcriptase can be added to the RPA reaction for amplification of RNA targets. All reagents are glycerol-free, which enables assay lyophilization.
Features of Lyo-ready RPA products
- Enhanced sensitivity down to 1 copy using optimized (RT-)RPA reaction buffer
- Robust and specific RNA/DNA amplification
- Amplification from inhibitor-containing samples
- Fast target amplification in as little as 20 minutes
- Ability to easily optimize RPA or RT-RPA reactions
Kit components
The Lyo-ready RPA Kit includes all required components for an RPA reaction:
- Lyo-ready T4 UvsX Protein—produced from E. coli cells bearing cloned uvsX gene derived from bacteriophage T4
- Lyo-ready T4 UvsY Protein—a recombination mediator protein that stimulates homologous recombination performed by the UvsX recombinase
- Lyo-ready Bst DNA Polymerase (Cat. No. A56655)—a proprietary mutant of Bst DNA Polymerase, Large Fragment, with fast reaction speed, increased sensitivity, and tolerance to inhibitors
- Lyo-ready T4 Gene 32 Protein—a single-stranded DNA binding protein produced from E. coli cells carrying cloned bacteriophage T4 gene 32
- Optimized reaction buffer, magnesium chloride, and dNTPs
Specifications
Specifications
| Product Type | Reaction Buffer |
| Form | Liquid |
| For Use With (Application) | RPA, RT-RPA |
| Quantity | 2000 μL |
| Research Category | Nucleic Acid Amplification |
| Content And Storage | 2X RPA Reaction Buffer, 2 mL, Store at -25°C to -15°C. |
| Optimal Reaction Temperature | 42°C |
| Shipping Condition | Dry Ice |
| Concentration | 2X |
| Reaction Time | Up to 20 min. |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.